missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA7 Antibody (6C8G7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 222.00
Specifications
| Antigen | EphA7 |
|---|---|
| Clone | 6C8G7 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
EphA7 Monoclonal specifically detects EphA7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EphA7 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
| EPHA7 | |
| IgG2b | |
| Unpurified |
| 6C8G7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
| Q15375 | |
| 2045 | |
| Purified recombinant fragment of EPH Receptor A7 (aa27-210) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title