missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphA8 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148865
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
Specifications
| EphA8 | |
| Polyclonal | |
| Unconjugated | |
| Epha8 | |
| AW047546; Eek; EK3; epa8; EPH- and ELK-related kinase; EPH- and ELK-related tyrosine kinase; Eph receptor A8; Epha8; EPH-like kinase 3; Ephrin type-A receptor 8; Ephrin type-A receptor 8 precursor (Tyrosine-protein kinase receptor EEK) (EPH-and ELK-related kinase); Hek3; hydroxyaryl-protein kinase; KIAA1459; mKIAA1459; protein-tyrosine kinase; tyrosine-protein kinase receptor EEK; tyrosylprotein kinase | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 13842 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O09127 | |
| Epha8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA8, Gly27 - Arg540. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction