missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB2 Antibody (512012), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB467L
This item is not returnable.
View return policy
Description
EphB2 Monoclonal antibody specifically detects EphB2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| EphB2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 512012 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CAPB, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, ERKHek5, HEK5, PCBC, protein-tyrosine kinase HEK5, Renal carcinoma antigen NY-REN-47, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB2, Val27-Lys548 (predicted), Accession # P54763 | |
| 0.1 mL | |
| Prostate Cancer, Protein Kinase | |
| 2048 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction