missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EphB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16353615
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16353615 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16353615 Supplier Invitrogen™ Supplier No. PA586749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

EPHB3 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. The ephrin ligands and Eph receptors display reciprocal expression in vivo. Developing and adult neural tissue express nearly all of the Eph receptors and ephrin ligands. Ephs and ephrins play a significant role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphB3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene EPHB3
Gene Accession No. P54753, P54754
Gene Alias AW456895; Cek10; Developmental kinase 5; EK2; Embryonic kinase 2; EPH receptor B3; EPHB3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin type-B receptor 3; Etk2; HEK2; human embryo kinase 2; Mdk5; mDK-5; Sek4; TYRO6; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6
Gene Symbols EPHB3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in Human EphB3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13845, 2049, 287989
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.