missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB4 Antibody (047), DyLight 550 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89353R
This item is not returnable.
View return policy
Description
EphB4 Monoclonal antibody specifically detects EphB4 in Human samples. It is validated for ELISA
Specifications
| EphB4 | |
| Monoclonal | |
| DyLight 550 | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB4 (NP_004435.3; Met1-Ala539). | |
| 0.1 mL | |
| Protein Kinase | |
| 2050 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 047 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction