missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB6 Antibody (026), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89326AF405
This item is not returnable.
View return policy
Description
EphB6 Monoclonal antibody specifically detects EphB6 in Human samples. It is validated for ELISA, ELISA
Specifications
| EphB6 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 026 | |
| ELISA, Sandwich ELISA | |
| EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB6 (NP_004436.1; Met1-Ser579). | |
| 0.1 mL | |
| Protein Kinase | |
| 2051 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction