Learn More
EphB6 Antibody (465327) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB33841L
Product Code. 18670750
  • € 597.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
EphB6 Monoclonal antibody specifically detects EphB6 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EphB6
Monoclonal
DyLight 550
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
465327
Flow Cytometry, CyTOF-ready
EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911
Mouse myeloma cell line NS0-derived recombinant human EphB6, Val17-Thr579, Accession # O15197
0.1 mL
Protein Kinase
2051
Store at 4°C in the dark.