Learn More
Novus Biologicals™ EphB6 Antibody Pair [HRP]
Product Code. 18212696 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. 18212696
Brand: Novus Biologicals™ NBP27949015PLATES
Rabbit Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human EphB6.
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 9.4 pg/mL
Assay Sensitivity 9.38-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human EphB6.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911
Gene ID (Entrez) 2051
Gene Symbol EPHB6
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 8 ng of recombinant EphB6.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-EphB6 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-EphB6 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Protein Kinase
    Storage Requirements Storage is content dependent.
    Target EphB6
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.6 ng/mL is recommended.