missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A1 Antibody (067), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89659CL1
This item is not returnable.
View return policy
Description
Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Rat samples. It is validated for ELISA
Specifications
| Ephrin-A1 | |
| Monoclonal | |
| CoraFluor 1 | |
| ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Rat Ephrin-A1 (Uniprot#: P97553; Met1-His181). | |
| 0.1 mL | |
| Cancer | |
| 1942 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 067 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction