missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A1 Antibody (RM0224-6D31), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12277AF750
This item is not returnable.
View return policy
Description
Ephrin-A1 Monoclonal specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blot.
Specifications
| Ephrin-A1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EFNA1 | |
| Mouse recombinant Ephrin-A1 | |
| 0.1 mL | |
| 1942 | |
| Mouse | |
| IgG2 |
| Western Blot | |
| RM0224-6D31 | |
| Western Blot | |
| ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Detects mouse Ephrin-A1 with WB and shows weaker cross-reactivity with mouse Ephrin-A2. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction