missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A2 Antibody (OTI3E3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70622H
This item is not returnable.
View return policy
Description
Ephrin-A2 Monoclonal specifically detects Ephrin-A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Ephrin-A2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| EFNA2 | |
| Full length recombinant protein of human Ephrin-A2 (NP_001396) produced in HEK293T cells. | |
| 0.1 mL | |
| 1943.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3E3 | |
| Western Blot, Immunohistochemistry | |
| ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6LERK-6, ligand of eph-related kinase 6 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction