missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A3 Antibody (OTI4A4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70623H
This item is not returnable.
View return policy
Description
Ephrin-A3 Monoclonal specifically detects Ephrin-A3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| Ephrin-A3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| EFNA3 | |
| Recombinant protein fragment corresponding to amino acids 23-214 of human Ephrin-A3 (NP_004943) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Immunohistochemistry | |
| OTI4A4 | |
| Immunohistochemistry | |
| EFL2, EFL-2, EHK1-L, EPH-related receptor tyrosine kinase ligand 3, ephrin-A3, EPLG3EHK1 ligand, LERK3LERK-3, ligand of eph-related kinase 3 | |
| Mouse | |
| Affinity Purified | |
| Growth and Development, Neuronal Cell Markers | |
| 1944.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction