Learn More
Novus Biologicals™ Ephrin-A4 Antibody (PF-06647263) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28626-100ug
This item is not returnable.
View return policy
Description
Ephrin-A4 Monoclonal antibody specifically detects Ephrin-A4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Ephrin-A4 | |
| Monoclonal | |
| Unconjugated | |
| P52798 | |
| Human | |
| Protein A purified | |
| RUO | |
| 1945 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| PF-06647263 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4 | |
| EFNA4 | |
| 100 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |