Learn More
Novus Biologicals™ Ephrin-A4 Antibody (PF-06647263) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28626-100ug
 View more versions of this product
Product Code. 30046656
  • € 349.00 / 100µg
Description
Ephrin-A4 Monoclonal antibody specifically detects Ephrin-A4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ephrin-A4
Monoclonal
Unconjugated
P52798
Human
Protein A purified
RUO
1945
Human
Purified
Flow Cytometry, ELISA, Functional Assay
PF-06647263
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4
EFNA4
100 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.