missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ephrin A4 Monoclonal Antibody (132416)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561582

Product Code. 30535855

  • € 589.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Ephrin A4 is also known as LERK-4 or ELF-4. Ephrin-A4 binds EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphB1. To date, at least 14 members of the Eph receptor family and a family of 8 ligands have been identified. Ligands of Eph family receptors are structurally related membrane-bound proteins that can be subdivided into two major subclasses: ephrin-A and ephrin-B. Ligands in the ephrin-A subclass, including the prototype family member ephrin-A1 (B61), are membrane associated through glycosylphosphatidyl-inositollinkages; whereas ephrin-B subclass consists of ligands with transmembrane domains.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ephrin A4
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O08542
EFNA4
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Ephrin-A4, Leu26-Gly176.
500 μg
Primary
Mouse
Antibody
IgG2b
Western Blot
132416
Unconjugated
EFNA4
EFL4; EFL-4; Efna4; EPH-related receptor tyrosine kinase ligand 4; ephrin A4; ephrin-A4; Epl4; EPLG4; LERK4; LERK-4; ligand of eph-related kinase 4
Rat
Protein A/G
RUO
13639
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.