missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-A5 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98318AF405
This item is not returnable.
View return policy
Description
Ephrin-A5 Polyclonal antibody specifically detects Ephrin-A5 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Ephrin-A5 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| AF1, AL-1, EPH-related receptor tyrosine kinase ligand 7, ephrin-A5, EPLG7, GLC1M, LERK-7, LERK7EFL5, RAGS | |
| Produced in rabbits immunized with purified, recombinant Mouse Ephrin-A5 (Uniprot#: O08543-1; Met1-Asn203) | |
| 0.1 mL | |
| Neuroscience, Vision | |
| 1946 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction