missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin-B2 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98321C
This item is not returnable.
View return policy
Description
Ephrin-B2 Polyclonal specifically detects Ephrin-B2 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ephrin-B2 | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ephrin B2, EPLG5, Htk-L, HTKL, LERK5 | |
| Produced in rabbits immunized with purified, recombinant Ephrin-B2 (NP_034241.2; Met1-Ala232). | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 1948 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction