missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHX1 Antibody (OTI3F10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70646V
This item is not returnable.
View return policy
Description
EPHX1 Monoclonal specifically detects EPHX1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EPHX1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EPHX1 | |
| Recombinant protein fragment corresponding to amino acids 21-230 of human EPHX1 (NP_000111) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI3F10 | |
| Western Blot, Immunohistochemistry | |
| EC 3.3.2.9, EPHX, EPOX, Epoxide hydratase, epoxide hydrolase 1, epoxide hydrolase 1, microsomal (xenobiotic), MEH, Microsomal epoxide hydrolase | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 2052.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction