missing translation for 'onlineSavingsMsg'
Learn More

EPHX2 Antibody (OTI1A6), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-70648B

Product Code. 18109302

  • ONLINE EXCLUSIVE PRICE
    € 785.00
    € 742.35 / 0.10mL
    Save € 42.65
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EPHX2 Monoclonal specifically detects EPHX2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EPHX2
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
EPHX2
Full length recombinant protein of human EPHX2 (NP_001970) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1A6
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CEH, Cytosolic epoxide hydrolase, EC 3.3.2.10, Epoxide hydratase, epoxide hydrolase 2, epoxide hydrolase 2, cytoplasmic, epoxide hydrolase 2, cytosolic, epoxide hydrolase, soluble, SEH, Soluble epoxide hydrolase
Mouse
Affinity Purified
Lipid and Metabolism
2053.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.