missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHX2 Monoclonal Antibody (OTI3F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501628S
This item is not returnable.
View return policy
Description
EPHX2 Monoclonal antibody specifically detects EPHX2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| EPHX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P34913 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2053 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI3F6 | |
| Unconjugated | |
| EPHX2 | |
| bifunctional epoxide hydrolase 2; CEH; cytosolic epoxide hydrolase; Cytosolic epoxide hydrolase 2; Eph2; Ephx2; Epoxide hydratase; epoxide hydrolase 2; epoxide hydrolase 2, cytoplasmic; epoxide hydrolase 2, cytosolic; epoxide hydrolase 2C; epoxide hydrolase, soluble; Lipid-phosphate phosphatase; SEH; sEP; Soluble epoxide hydrolase | |
| Full length human recombinant protein of human EPHX2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction