missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EPHX2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533517

Product Code. 15304791

  • € 530.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Orangutan (95%) Gibbon, Monkey (89%) Marmoset (84%).

This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EPHX2
Polyclonal
Unconjugated
EPHX2
bifunctional epoxide hydrolase 2; CEH; cytosolic epoxide hydrolase; Cytosolic epoxide hydrolase 2; Eph2; Ephx2; Epoxide hydratase; epoxide hydrolase 2; epoxide hydrolase 2, cytoplasmic; epoxide hydrolase 2, cytosolic; epoxide hydrolase 2C; epoxide hydrolase, soluble; Lipid-phosphate phosphatase; SEH; sEP; Soluble epoxide hydrolase
Rabbit
Antigen affinity chromatography
RUO
2053
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
P34913
EPHX2
Synthetic 19 amino acid peptide from Near the N-terminus of human EPHX2.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.