Learn More
(-)-Epicatechin, MedChemExpress
Product Code. 16464717
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464717 50 mg 50mg
16418818 10 mg 10mg
16488946 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16464717 Supplier Medchem Express Supplier No. HYN0001
MedChemExpress (-)-Epicatechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 3.2 μM. (-)-Epicatechin inhibits the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB.
(-)-Epicatechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 3.2 μM. (-)-Epicatechin inhibits the IL-1β-induced expression of iNOS by blocking the nuclear localization of the p65 subunit of NF-κB.
Specifications
Chemical Name or Material (-)-Epicatechin
CAS 490-46-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H14O6
Quantity 50 mg
Synonym (-)-Epicatechol Epicatechin epi-Catechin
Solubility Information DMSO : 100 mg/mL (344.51 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1
Molecular Weight (g/mol) 290.27
Formula Weight 290.27
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only