missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epigen Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA579209
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A431 whole cell, rat PC-12 whole cell.
Epigen is an EGF-related polypeptide growth factor that signals through the ErbB receptor-1. Promotes the growth of epithelial cells. May stimulate the phosphorylation of EGFR and mitogen-activated protein kinases. It is produced in several tissues, including the testis, liver, and heart, as well as in certain tumor cells. Epigen is mitogenic for fibroblasts and epithelial cells. Human Epigen is initially synthesized as a glycosylated 14.7 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a mature soluble sequence.
Especificaciones
| Epigen | |
| Polyclonal | |
| Unconjugated | |
| EPGN | |
| 2310069M11Rik; ALGV3072; EPG; EPGN; epigen; epithelial mitogen; epithelial mitogen homolog; PRO9904; RGD1560084; UNQ3072/PRO9904 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 255324, 289515 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| Q6UW88 | |
| EPGN | |
| E. coli-derived human Epigen recombinant protein (Position: A23-K110). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido