missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epiregulin Antibody (183629), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB14251R
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal antibody specifically detects Epiregulin in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Epiregulin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 183629 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| epiregulin, ER, proepiregulin | |
| E. coli-derived recombinant human Epiregulin, Val63-Leu108, Accession # O14944 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins | |
| 2069 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction