missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epiregulin Antibody (183629) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB14251B
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal antibody specifically detects Epiregulin in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Epiregulin | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 183629 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| epiregulin, ER, proepiregulin | |
| E. coli-derived recombinant human Epiregulin, Val63-Leu108, Accession # O14944 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins | |
| 2069 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction