missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epiregulin Antibody (MM0265- 12Y4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12285FR
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Epiregulin | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EREG | |
| Human recombinant Epiregulin | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0265- 12Y4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| epiregulin, ER, proepiregulin | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Biologically Active Proteins | |
| 2069 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction