missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelia (colonic) Antibody (PR 4D1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2689AF350
This item is not returnable.
View return policy
Description
Epithelia (colonic) Monoclonal specifically detects Epithelia (colonic) in Human samples. It is validated for Immunohistochemistry.
Specifications
| Epithelia (colonic) | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mucosal scrapings and membrane preparations from normal colorectal epithelium and possibly booster inoculations with the colon carcinoma cell line HT29. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| PR 4D1 | |
| Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| Cancer, Cellular Markers, Extracellular Matrix | |
| This antibody has wide reactivity with GI tract, and reacts with a high MW glycoprotein in human colonic mucus, epithelial cells and cell membranes. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction