missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelia (colonic) Antibody (PR 4D1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB100-2689F
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Epithelia (colonic) Monoclonal specifically detects Epithelia (colonic) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Spezifikation
| Epithelia (colonic) | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MUC1 | |
| Mucosal scrapings and membrane preparations from normal colorectal epithelium and possibly booster inoculations with the colon carcinoma cell line HT29. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix | |
| 4582 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| PR 4D1 | |
| Immunohistochemistry | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody has wide reactivity with GI tract, and reacts with a high MW glycoprotein in human colonic mucus, epithelial cells and cell membranes. | |
| Store at 4C in the dark. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur