missing translation for 'onlineSavingsMsg'
Learn More

epithelial Sodium Channel alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 386.00 - € 529.00

Specifications

Antigen epithelial Sodium Channel alpha
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18477980
View Documents
Novus Biologicals
NBP1-84846-25ul
25 μL
€ 386.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18433301
View Documents
Novus Biologicals
NBP1-84846
0.1 mL
€ 529.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

epithelial Sodium Channel alpha Polyclonal antibody specifically detects epithelial Sodium Channel alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

epithelial Sodium Channel alpha
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Phospho Specific
PBS (pH 7.2) and 40% Glycerol
6337
IgG
Immunogen affinity purified
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Purified
RUO
Human
alpha ENaC-2, alpha-ENaC, Alpha-NaCH, amiloride-sensitive epithelial sodium channel alpha subunit, amiloride-sensitive sodium channel subunit alpha, amiloride-sensitive sodium channel subunit alpha 2, BESC2, ENaCA, ENaCalpha, Epithelial Na(+) channel subunit alpha, FLJ21883, nasal epithelial sodium channel alpha subunit, Nonvoltage-gated sodium channel 1 subunit alpha, SCNEA, SCNN1, sodium channel, nonvoltage-gated 1 alpha
This antibody was developed against Recombinant Protein corresponding to amino acids: RYPEIKEELEELDRITEQTLFDLYKYSSFTTLVAGSRSRRDLRGTLPHPLQRLRVPPPPHGARRARSVASSLRDNNPQVDWKDWKIGFQL
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.