missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epithelial Sodium Channel gamma Antibody (11-35-1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41374JF646
This item is not returnable.
View return policy
Description
epithelial Sodium Channel gamma Monoclonal antibody specifically detects epithelial Sodium Channel gamma in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| epithelial Sodium Channel gamma | |
| Monoclonal | |
| Janelia Fluor 646 | |
| amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma | |
| Synthetic peptide seguence: VGGSIIHKAC | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 11-35-1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6340 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction