Learn More
epithelial Sodium Channel gamma Antibody (5c2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-41373AF700
Product Code. 18700432
  • € 746.00 / 100µL
Description
epithelial Sodium Channel gamma Monoclonal antibody specifically detects epithelial Sodium Channel gamma in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
epithelial Sodium Channel gamma
Monoclonal
Alexa Fluor 700
amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma
The inhibitory peptide from the human yENaC subunit. EAESWNSVSEGKQPRFSHRIPLC corresponding to amino acid residue 139-160 of human yENaC subunit.
100 μg
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
5c2
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
6340
Store at 4°C in the dark.
IgG1 κ