missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epithelial Sodium Channel gamma Antibody (5c2), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41373V
This item is not returnable.
View return policy
Description
epithelial Sodium Channel gamma Monoclonal antibody specifically detects epithelial Sodium Channel gamma in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Sandwich ELISA
Specifications
| epithelial Sodium Channel gamma | |
| Monoclonal | |
| DyLight 405 | |
| amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma | |
| The inhibitory peptide from the human yENaC subunit. EAESWNSVSEGKQPRFSHRIPLC corresponding to amino acid residue 139-160 of human yENaC subunit. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 5c2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6340 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction