missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPLIN Antibody (OTI1A3), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71932JF549
This item is not returnable.
View return policy
Description
EPLIN Monoclonal specifically detects EPLIN in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EPLIN | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LIMA1 | |
| Recombinant protein fragment corresponding to amino acids 1-275 of human EPLIN (NP_057441) produced in E.coli. | |
| 0.1 mL | |
| 51474.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1A3 | |
| Western Blot, Immunohistochemistry | |
| 1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction