Learn More
EPM2AIP1 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18455JF585
This item is not returnable.
View return policy
Description
EPM2AIP1 Polyclonal antibody specifically detects EPM2AIP1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| EPM2AIP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9852 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| EPM2A (laforin) interacting protein 1, EPM2A interacting protein 1, EPM2A-interacting protein 1, KIAA0766FLJ11207, Laforin-interacting protein | |
| Recognizes region between residues Between Between 100 and 150.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |