Learn More
EPM2AIP1 Mouse anti-Human, Clone: OTI2A2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501790
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless.Specifications
| EPM2AIP1 | |
| Monoclonal | |
| Unconjugated | |
| EPM2AIP1 | |
| EPM2A interacting protein 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9852 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q7L775 | |
| EPM2AIP1 | |
| Full length human recombit protein of human EPM2AIP1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.