missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EpoR Monoclonal Antibody (38407)

Description
In sandwich ELISAs, no cross-reactivity or interference with recombinant human (rh) Epo, recombinant mouse Epo R, or rhThrombopoietin R was observed. Reconstitute at 0.5 mg/mL in sterile PBS.
EpoR encodes the erythropoietin receptor, which belongs to the cytokine receptor family. When erythropoietin binds to this receptor, it activates the Jak2 tyrosine kinase, which in turn activates various intracellular pathways including Ras/MAP kinase, phosphatidylinositol 3-kinase, and STAT transcription factors. The stimulated erythropoietin receptor plays a role in the survival of erythroid cells. Defects in this gene may result in erythroleukemia and familial erythrocytosis. Dysregulation of this gene may also affect the growth of certain tumors. Multiple transcript variants are produced as a result of alternate splicing. Diseases associated with EPOR include Erythrocytosis, Familial, 1 and Erythroleukemia.
Specifications
Specifications
| Antigen | EpoR |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 38407 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | EPOR |
| Gene Accession No. | P19235 |
| Gene Alias | EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?