missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EpoR Recombinant Rabbit Monoclonal Antibody (010), HRP

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552237
This item is not returnable.
View return policy
Description
EpoR Recombinant Monoclonal Antibody for ELISA
EpoR encodes the erythropoietin receptor, which belongs to the cytokine receptor family. When erythropoietin binds to this receptor, it activates the Jak2 tyrosine kinase, which in turn activates various intracellular pathways including Ras/MAP kinase, phosphatidylinositol 3-kinase, and STAT transcription factors. The stimulated erythropoietin receptor plays a role in the survival of erythroid cells. Defects in this gene may result in erythroleukemia and familial erythrocytosis. Dysregulation of this gene may also affect the growth of certain tumors. Multiple transcript variants are produced as a result of alternate splicing. Diseases associated with EPOR include Erythrocytosis, Familial, 1 and Erythroleukemia.
Specifications
| EpoR | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with no preservative | |
| P19235 | |
| EPOR | |
| Recombinant Human EPOR Protein (Ala 25-Pro250). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 010 | |
| HRP | |
| EPOR | |
| EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction