Learn More
Invitrogen™ EpoR Recombinant Rabbit Monoclonal Antibody (016)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA529248
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human EPOR/Erythropoietin Receptor.
EpoR encodes the erythropoietin receptor, which belongs to the cytokine receptor family. When erythropoietin binds to this receptor, it activates the Jak2 tyrosine kinase, which in turn activates various intracellular pathways including Ras/MAP kinase, phosphatidylinositol 3-kinase, and STAT transcription factors. The stimulated erythropoietin receptor plays a role in the survival of erythroid cells. Defects in this gene may result in erythroleukemia and familial erythrocytosis. Dysregulation of this gene may also affect the growth of certain tumors. Multiple transcript variants are produced as a result of alternate splicing. Diseases associated with EPOR include Erythrocytosis, Familial, 1 and Erythroleukemia.
Specifications
| EpoR | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7 | |
| P19235 | |
| EPOR | |
| Recombinant Human EPOR/Erythropoietin Receptor protein (Met1-Pro250). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 016 | |
| Unconjugated | |
| EPOR | |
| EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.