Learn More
Eppendorf™ S-4-104 Rotor Adapters
Product Code. 15188364 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1 x 250 mL Tubes
1 x 500 mL Bottle
14 x 15 mL Conical Tubes
14 x 5 mL Conical Tubes
20 x 5.5/12 mL Tubes
23 x 2.6/8 mL Tubes
23 x 7/17 mL Tubes
4 x 750 mL Wide-mouth Bottles
5 x 50 mL Conical Tubes
50 x 1.5/2 mL Tubes
7 x 50 mL Conical Tubes
Unit Size:
1 set
Pack of 2
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15188364 14 x 5 mL Conical Tubes Pack of 2
13256619 5 x 50 mL Conical Tubes Pack of 2
13236629 7 x 50 mL Conical Tubes Pack of 2
12886243 14 x 15 mL Conical Tubes Pack of 2
13276619 20 x 5.5/12 mL Tubes Pack of 2
13286619 23 x 2.6/8 mL Tubes Pack of 2
13216629 1 x 250 mL Tubes Pack of 2
13296619 50 x 1.5/2 mL Tubes Pack of 2
13206629 23 x 7/17 mL Tubes Pack of 2
13226629 4 x 750 mL Wide-mouth Bottles Pack of 2
15159043 1 x 500 mL Bottle 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15188364 Supplier Eppendorf™ Supplier No. 5825739000
Only null left
Add to basket
Add to basket
Includes
2 piece set
Available in a wide range of configurations to suit a variety of applications.
  • For use with Centrifuge 5810/5810 R
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Capacity 14 x 5 mL Conical Tubes
For Use With (Equipment) S-4-104 rotor for 5810/5810R centrifuge
No. per Pack 2 Pack