Learn More
Eppendorf Eppendorf twin.tec™ PCR plate 96, semi-skirted
Made from polypropylene, Eppendorf twin.tec PCR Plates LoBind enable you to recover nearly 100 % of your DNA/RNA molecules, and as there’s no surface coating, there’s no risk of sample contamination. Batch-tested and certified free from DNA, DNase, RNase and PCR inhibitors, they are perfect for PCR.
Brand: Eppendorf 0030129695
Additional Details : Weight : 0.42000kg
Description
Eppendorf twin.tec™ PCR plate 96, semi-skirtedSpecifications
10 |