missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPRS Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22311IR
This item is not returnable.
View return policy
Description
EPRS Polyclonal antibody specifically detects EPRS in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EPRS | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| bifunctional aminoacyl-tRNA synthetase, Cell proliferation-inducing gene 32 protein, DKFZp313B047, EARS, EC 6.1.1, EC 6.1.1.15, EC 6.1.1.18, GLNS, GLUPRORS, glutamate tRNA ligase, glutaminyl-tRNA synthetase, glutamyl-prolyl-tRNA synthetase, PARSproliferation-inducing gene 32 protein, proliferation-inducing protein 32, proline tRNA ligase, proline-tRNA ligase, prolyl-tRNA synthetase, QARS, QPRS | |
| The immunogen this antibody was made to, maps to a region between residue 950 to 1000 of human Glutamyl-prolyl-tRNA Synthetase using the numbering given in entry NP_004437.2 (GeneID 2058). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 2058 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction