missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eps8 Antibody (18F5B11), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27344AF532
This item is not returnable.
View return policy
Description
Eps8 Monoclonal specifically detects Eps8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Eps8 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EPS8 | |
| Amino acids 701-882 of human EPS8 protein were used as the immunogen for the EPS8 antibody. | |
| Primary | |
| Human | |
| IgG3 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 18F5B11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| epidermal growth factor receptor kinase substrate 8, epidermal growth factor receptor pathway substrate 8 | |
| Mouse | |
| 0.1 mL | |
| 2059 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction