missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS8L3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98054AF350
This item is not returnable.
View return policy
Description
EPS8L3 Polyclonal specifically detects EPS8L3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| EPS8L3 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| epidermal growth factor receptor kinase substrate 8-like protein 3, Epidermal growth factor receptor pathway substrate 8-related protein 3, EPS8-like 3, EPS8-like protein 3, EPS8R3, EPS8-related protein 3, FLJ21522, MGC16817 | |
| Produced in rabbits immunized with E. coli-derived Human EPS8L3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 79574 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction