Learn More
Epsilon-n-methyl-l-lysine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Epsilon-n-methyl-l-lysine Hydrochloride |
| CAS | 7622-29-9 |
| Molecular Formula | C7H16N2O2 • HCl |
| Quantity | 100 mg |
| Synonym | Epsilon-N-Methyl-L-Lysine Hcl |
| InChI Formula | InChI=1S/C7H16N2O2.ClH/c1-9-5-3-2-4-6(8)7(10)11;/h6,9H,2-5,8H2,1H3,(H,10,11);1H/t6-;/m0./s1 |
| SMILES | N[C@@H](CCCCNC)C(O)=O.Cl |
| IUPAC Name | (2S)-2-amino-6-(methylamino)hexanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 160.213646 |
| Formula Weight | 196.0979 |