missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epsin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 184.00 - € 485.00
Specifications
| Antigen | Epsin 1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17700514
![]() |
Invitrogen™
PA5119098 |
200 μL |
€ 485.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17703414
|
Invitrogen™
PA5122012 |
20 μL |
€ 184.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epsin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the epsin protein family. The encoded protein binds clathrin and is involved in the endocytosis of clathrin-coated vesicles. Loss of function of this gene is associated with reduced tumor growth and progression in certain cancer types.Specifications
| Epsin 1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| EPN1 | |
| AA269831; EH domain-binding mitotic phosphoprotein; Epn1; EPS-15-interacting protein 1; Epsin 1; epsin-1; hypothetical protein LOC508199; Ibp1; Intersectin-EH-binding protein 1; LOW QUALITY PROTEIN: epsin-1 | |
| EPN1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q80VP1, Q9Y6I3 | |
| 13854, 29924 | |
| Recombinant protein Epsin 1. The antigen corresponds to amino acid range 279-437 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title