missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epsin-2 Antibody (OTI1G3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71701AF532
This item is not returnable.
View return policy
Description
Epsin-2 Monoclonal specifically detects Epsin-2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Epsin-2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EPN2 | |
| Full length recombinant protein of human Epsin-2 (NP_055779) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1G3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Eps15 binding protein, EPS-15-interacting protein 2, epsin 2, epsin-2, KIAA1065EHB21 | |
| Mouse | |
| 0.1 mL | |
| 22905 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction