missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epsin 3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98974JF549
This item is not returnable.
View return policy
Description
Epsin 3 Polyclonal antibody specifically detects Epsin 3 in Human samples. It is validated for Western Blot
Specifications
| Antigen and protein A Affinity-purified | |
| 55040 | |
| Store at 4C in the dark. |
| 0.1 mL | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction