missing translation for 'onlineSavingsMsg'
Learn More

Epstein Barr Virus Antibody (CS1-4), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-08501CL1

Product Code. 30150769

  • € 1165.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Epstein Barr Virus Monoclonal antibody specifically detects Epstein Barr Virus in Virus samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epstein Barr Virus
Monoclonal
CoraFluor 1
Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1
Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP
0.1 mL
Primary
Store at 4°C in the dark. Do not freeze.
IgG1 κ
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CS1-4
PBS
Mouse
Protein A or G purified
RUO
Virus
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.