missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPSTI1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145423
This item is not returnable.
View return policy
Description
EPSTI1 was up-regulated in breast carcinomas. The exact function of EPSTI1 is not known.
Specifications
| EPSTI1 | |
| Polyclonal | |
| Unconjugated | |
| EPSTI1 | |
| 2310046K10Rik; 5033415K03Rik; BRESI1; epithelial stromal interaction 1; epithelial stromal interaction 1 (breast); epithelial-stromal interaction protein 1; EPSTI1; RGD1563207; Unknown (protein for IMAGE:7987136) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100625050, 94240 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q96J88 | |
| EPSTI1 | |
| Recombinant Human EPSTI1 protein, Asn73-Glu280 (Accession #Q96J88). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction