Learn More
MedChemExpress Eptifibatide
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Purity | 96.49% |
| CAS | 188627-80-7 |
| Content And Storage | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 831.96 g/mol |
| Product Type | Peptides |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 250 mg/mL (300.50 mM∣Need ultrasonic) ∣H2O : 50 mg/mL (60.10 mM∣Need ultrasonic) |