missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPX Antibody (rEPO104), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08457AF750
This item is not returnable.
View return policy
Description
EPX Monoclonal antibody specifically detects EPX in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| EPX | |
| Monoclonal | |
| Alexa Fluor 750 | |
| EC 1.11.1.7, eosinophil peroxidase, EPOEPER, EPPEC 1.11.1, EPX-PEN | |
| Human eosinophils from a patient with hypereosinophilic syndrome (Uniprot: P11678 ) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| rEPO104 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 8288 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction